siRNA Details
| Gene Symbol: Stam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019667.2 | NP_062641.1 | 14 | 1538 | ||
| GenBank mRNA | AB012611.1 | - | 14 | 1553 | ||
| AK004604.2 | - | 14 | 1538 | |||
| AK035789.1 | - | 1591 | ||||
| AK159699.1 | - | 14 | 1567 | |||
| AK168729.1 | - | 14 | 1570 | |||
| BC013818.1 | - | 14 | 1574 | 3671426 | ||

Stam2
Genomic Map