siRNA Details
| Gene Symbol: Dnajc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019795.4 | NP_062769.2 | 2 | 667 | ||
| GenBank mRNA | AB028861.1 | - | 2 | 115 | ||
| AK011875.1 | - | 2 | 106 | |||
| AK076032.1 | - | 2 | 138 | |||
| AK145506.1 | - | 2 | 129 | |||
| AK145739.1 | - | 2 | 129 | |||
| AK166872.1 | - | 2 | 129 | |||
| AY323828.1 | - | 2 | 103 | |||
| BC023681.1 | - | 2 | 311 | 5343384 | ||
| BC055729.1 | - | 2 | 666 | 6414499 | ||

Dnajc7
Genomic Map