siRNA Details
| Gene Symbol: Fzr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019757.1 | NP_062731.1 | 7 | 693 | ||
| GenBank mRNA | AF083809.1 | - | 7 | 693 | ||
| AK051993.1 | - | 7 | 776 | |||
| AK083686.1 | - | 756 | ||||
| AK154822.1 | - | 7 | 777 | |||
| AK155370.1 | - | 7 | 775 | |||
| AK163951.1 | - | 7 | 752 | |||
| AK172679.1 | - | 7 | 787 | |||
| AK203764.1 | - | 2 | 120 | |||
| BC006616.1 | - | 7 | 774 | 3967009 | ||

Fzr1
Genomic Map