siRNA Details
| Gene Symbol: Dusp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019819.3 | NP_062793.2 | 3 | 814 | ||
| GenBank mRNA | AF120113.1 | - | 1 | 540 | ||
| AK009744.1 | - | 3 | 827 | |||
| AK079362.1 | - | 3 | 815 | |||
| AK166230.1 | - | 3 | 812 | |||
| AK166840.1 | - | 3 | 810 | |||
| BC002130.1 | - | 3 | 779 | 3592420 | ||
| BC052836.1 | - | 3 | 834 | 30049007 | ||

Dusp14
Genomic Map