siRNA Details
Gene Symbol: Trpc4ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163452.1 | NP_001156924.1 | 2, 3 | 376 | ||
NM_019828.2 | NP_062802.2 | 2, 3 | 376 | |||
GenBank mRNA | AF130458.1 | - | 2, 3 | 324 | ||
AF509881.1 | - | 2, 3 | 298 | |||
AK076496.1 | - | 2, 3 | 375 | |||
AK140799.1 | - | 1, 2 | 64 | |||
AK154263.1 | - | 2, 3 | 325 | |||
AK156047.1 | - | 2, 3 | 325 | |||
AK165259.1 | - | 2, 3 | 325 | |||
AK170085.1 | - | 2, 3 | 314 | |||
AK171404.1 | - | 2, 3 | 325 | |||
AK178651.1 | - | 1, 2 | 142 | |||
AK202675.1 | - | 1, 2 | 142 | |||
AK216771.1 | - | 2, 3 | 193 | |||
AK220239.1 | - | 2, 3 | 352 | |||
BC033274.1 | - | 2, 3 | 304 | 3673725 | ||
BC057330.1 | - | 2, 3 | 334 | 6816155 |
