siRNA Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019670.1 | NP_062644.1 | 20 | 2243 | ||
| GenBank mRNA | AF094519.1 | - | 20 | 2243 | ||
| AK138694.1 | - | 20 | 2273 | |||
| AK161737.1 | - | 19 | 2261 | |||
| AK166882.1 | - | 20 | 2299 | |||
| AK220493.1 | - | 20 | 2307 | |||
| BC028920.1 | - | 13 | 1544 | 4008236 | ||
| BC085191.1 | - | 20 | 2353 | 30693873 | ||
| BC086779.1 | - | 20 | 2318 | |||

Diap3
Genomic Map