siRNA Details
| Gene Symbol: Dstn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019771.2 | NP_062745.1 | 2, 3 | 621 | ||
| GenBank mRNA | AB025406.1 | - | 2, 3 | 341 | ||
| AK078898.1 | - | 2, 3 | 395 | |||
| BC131926.1 | - | 2, 3 | 338 | 40130203 | ||
| CT010216.1 | - | 1, 2 | 307 | |||

Dstn
Genomic Map