siRNA Details
| Gene Symbol: Dstn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019771.2 | NP_062745.1 | 4 | 743 | ||
| GenBank mRNA | AB025406.1 | - | 4 | 463 | ||
| AK078898.1 | - | 4 | 517 | |||
| BC131926.1 | - | 4 | 460 | 40130203 | ||
| CT010216.1 | - | 3 | 429 | |||

Dstn
Genomic Map