siRNA Details
| Gene Symbol: Dstn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019771.2 | NP_062745.1 | 2 | 375 | ||
| GenBank mRNA | AB025406.1 | - | 2 | 95 | ||
| AK078898.1 | - | 2 | 149 | |||
| BC131926.1 | - | 2 | 92 | 40130203 | ||
| BF123977.1 | - | 2 | 386 | |||
| CT010216.1 | - | 1 | 61 | |||

Dstn
Genomic Map