siRNA Details
| Gene Symbol: Ctsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019861.1 | NP_063914.1 | 10 | 1164 | ||
| GenBank mRNA | AF136280.1 | - | 10 | 1065 | ||
| AF197480.1 | - | 10 | 1109 | |||
| AJ131851.1 | - | 10 | 1041 | |||
| AK017474.1 | - | 10 | 1047 | |||
| AK075862.1 | - | 10 | 1057 | |||
| AK207816.1 | - | 3 | 183 | |||
| BC004054.1 | - | 7 | 635 | |||
| BC051131.1 | - | 9 | 1283 | |||
| BC058758.1 | - | 10 | 1055 | 6492834 | ||
| BC115438.1 | - | 4 | 284 | |||

Ctsf
Genomic Map