siRNA Details
| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019741.3 | NP_062715.2 | 7, 8 | 1220 | ||
| GenBank mRNA | AF161071.1 | - | 5, 6 | 581 | ||
| AK029720.1 | - | 7, 8 | 1221 | |||
| BC023500.1 | - | 5, 6 | 596 | 5043284 | ||

Slc2a5
Genomic Map