siRNA Details
| Gene Symbol: Vmn2r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019918.2 | NP_064302.2 | 3 | 838 | ||
| GenBank mRNA | AB267725.1 | - | 3 | 838 | ||
| AF053986.1 | - | 3 | 838 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r1| Gene Symbol: Vmn2r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019918.2 | NP_064302.2 | 3 | 838 | ||
| GenBank mRNA | AB267725.1 | - | 3 | 838 | ||
| AF053986.1 | - | 3 | 838 | |||
Set Membership: |
|