siRNA Details
| Gene Symbol: Rps6ka4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019924.2 | NP_064308.2 | 5 | 582 | ||
| GenBank mRNA | AF074714.1 | - | 5 | 554 | ||
| AK089467.1 | - | 5 | 573 | |||
| AK154674.1 | - | 5 | 581 | |||
| BC012964.1 | - | 5 | 592 | 4160873 | ||

Rps6ka4
Genomic Map