siRNA Details
| Gene Symbol: Gpr132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019925.4 | NP_064309.1 | 3 | 446 | ||
| GenBank mRNA | AF083442.1 | - | 3 | 482 | ||
| AK089866.1 | - | 3 | 423 | |||
| AK153765.1 | - | 3 | 447 | |||
| AK154509.1 | - | 3 | 468 | |||
| AK157600.1 | - | 3 | 474 | |||
| BC120522.1 | - | 2 | 373 | 40129445 | ||
| BC120524.1 | - | 2 | 373 | 40129447 | ||
More Information
Set Membership: |
|

Gpr132
Genomic Map