siRNA Details
| Gene Symbol: Hist1h1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020034.2 | NP_064418.1 | 1 | 311 | ||
| GenBank mRNA | AK020117.1 | - | 1 | 312 | ||
| BC114579.1 | - | 1 | 239 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h1b| Gene Symbol: Hist1h1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020034.2 | NP_064418.1 | 1 | 311 | ||
| GenBank mRNA | AK020117.1 | - | 1 | 312 | ||
| BC114579.1 | - | 1 | 239 | |||