siRNA Details
| Gene Symbol: Mlst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252463.1 | NP_001239392.1 | 5 | 498 | ||
| NM_001252464.1 | NP_001239393.1 | 5 | 473 | |||
| NM_001252465.1 | NP_001239394.1 | 3 | 282 | |||
| NM_019988.5 | NP_064372.2 | 5 | 493 | |||
| GenBank mRNA | AK002751.1 | - | 5 | 459 | ||
| AK038515.1 | - | 4 | 991 | |||
| AK040100.1 | - | 5 | 494 | |||
| AK048102.1 | - | 5 | 457 | |||
| AK077680.1 | - | 5 | 464 | |||
| AK083731.1 | - | 5 | 434 | |||
| AK088951.1 | - | 5 | 467 | |||
| BC015279.1 | - | 5 | 452 | 4222934 | ||
| BM947104.1 | - | 3 | 264 | |||
| CF750475.1 | - | 5 | 462 | |||

Mlst8
Genomic Map