siRNA Details
| Gene Symbol: Mllt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019914.4 | NP_064298.1 | 2 | 694 | ||
| GenBank mRNA | AB000733.1 | - | 2 | 268 | ||
| AK046005.1 | - | 2 | 199 | |||
| AK152708.1 | - | 2 | 694 | |||
| AK158495.1 | - | 2 | 320 | |||
| BC022963.1 | - | 2 | 135 | 5365628 | ||
| U95498.1 | - | 2 | 255 | |||

Mllt11
Genomic Map