siRNA Details
| Gene Symbol: Ralgapa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003719.2 | NP_001003719.1 | 28 | 4677 | ||
| NM_001112714.2 | NP_001106185.1 | 28 | 4677 | |||
| NM_001286263.1 | NP_001273192.1 | 29 | 4818 | |||
| NM_019994.5 | NP_064378.4 | 28 | 4677 | |||
| GenBank mRNA | AB032400.1 | - | 1 | 117 | ||
| AY066011.1 | - | 16 | 2263 | |||
| AY596972.1 | - | 28 | 4328 | |||
| AY596973.1 | - | 28 | 4328 | |||
| AY596974.1 | - | 28 | 4328 | |||
| BC145119.1 | - | 29 | 4399 | 9053667 | ||
| BC150707.1 | - | 29 | 4399 | 9087618 | ||

Ralgapa1
Genomic Map