siRNA Details
| Gene Symbol: Dnajb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159883.1 | NP_001153355.1 | 8 | 804 | ||
| NM_001159884.1 | NP_001153356.1 | 8 | 752 | |||
| NM_001159885.1 | NP_001153357.1 | 7 | 787 | |||
| NM_020266.2 | NP_064662.2 | 7 | 750 | |||
| NM_178055.4 | NP_835156.1 | 8 | 804 | |||
| GenBank mRNA | AB028858.1 | - | 7 | 617 | ||
| AK076061.1 | - | 8 | 740 | |||
| AK156083.1 | - | 8 | 773 | |||
| AK171494.1 | - | 8 | 750 | |||
| BC011090.1 | - | 8 | 706 | 3968468 | ||
| BC085162.1 | - | 4 | 3099 | |||

Dnajb2
Genomic Map