siRNA Details
| Gene Symbol: Aldh1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053080.3 | NP_444310.3 | 2 | 207 | ||
| GenBank mRNA | AF152359.1 | - | 2 | 162 | ||
| AF246711.1 | - | 2 | 155 | |||
| AF253409.1 | - | 2 | 194 | |||
| AF280404.1 | - | 2 | 159 | |||
| AK043530.1 | - | 2 | 204 | |||
| AK086764.1 | - | 2 | 220 | |||
| AK147006.1 | - | 2 | 208 | |||
| BC058277.1 | - | 2 | 151 | 6515355 | ||

Aldh1a3
Genomic Map