siRNA Details
| Gene Symbol: Slc37a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145960.1 | NP_001139432.1 | 5 | 752 | ||
| NM_020258.4 | NP_064654.3 | 5 | 752 | |||
| GenBank mRNA | AF121081.1 | - | 5 | 651 | ||
| AK028984.1 | - | 5 | 428 | |||
| AK031045.1 | - | 5 | 690 | |||
| AK079414.1 | - | 5 | 428 | |||
| AK079823.1 | - | 5 | 753 | |||
| AK220151.1 | - | 3 | 194 | |||
| BC038000.1 | - | 5 | 423 | |||
| BC063326.1 | - | 5 | 440 | 30301217 | ||

Slc37a2
Genomic Map