siRNA Details
| Gene Symbol: Slc37a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145960.1 | NP_001139432.1 | 5, 6 | 837 | ||
| NM_020258.4 | NP_064654.3 | 5, 6 | 837 | |||
| GenBank mRNA | AF121081.1 | - | 5, 6 | 736 | ||
| AK028984.1 | - | 5, 6 | 513 | |||
| AK031045.1 | - | 5, 6 | 775 | |||
| AK079414.1 | - | 5, 6 | 513 | |||
| AK079823.1 | - | 5, 6 | 838 | |||
| AK220151.1 | - | 3, 4 | 279 | |||
| BC038000.1 | - | 5, 6 | 508 | |||
| BC063326.1 | - | 5, 6 | 525 | 30301217 | ||

Slc37a2
Genomic Map