siRNA Details
| Gene Symbol: Zfp109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020262.3 | NP_064658.1 | 4 | 1770 | ||
| GenBank mRNA | AF167316.1 | - | 4 | 1732 | ||
| BC119619.1 | - | 3 | 1339 | 40062160 | ||
| BC119620.1 | - | 4 | 1397 | 40062167 | ||

Zfp109
Genomic Map