siRNA Details
| Gene Symbol: Pate4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020264.4 | NP_064660.2 | 1, 2 | 116 | ||
| GenBank mRNA | AF134204.1 | - | 1, 2 | 77 | ||
| AK020540.1 | - | 1, 2 | 100 | |||
| BC120764.1 | - | 1, 2 | 116 | 40129687 | ||
| BC120766.1 | - | 1, 2 | 116 | 40129689 | ||

Pate4
Genomic Map