siRNA Details
| Gene Symbol: Pdxp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020271.3 | NP_064667.2 | 2 | 874 | ||
| GenBank mRNA | AB041662.1 | - | 2 | 769 | ||
| AK043228.1 | - | 2 | 835 | |||
| AY366300.1 | - | 2 | 836 | |||
| BC037010.1 | - | 2 | 299 | |||
| BC046955.1 | - | 2 | 825 | |||
| BC049107.1 | - | 2 | 874 | |||
| BC058388.1 | - | 2 | 420 | |||
| BC069982.1 | - | 2 | 646 | |||
| BU148105.1 | - | 2 | 643 | |||

Pdxp
Genomic Map