siRNA Details
| Gene Symbol: Zfp276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020497.2 | NP_065243.2 | 5 | 1565 | ||
| GenBank mRNA | AF178935.1 | - | 4 | 607 | ||
| AK133363.1 | - | 5 | 1566 | |||
| BC138579.1 | - | 4 | 557 | 8861599 | ||
| BC138580.1 | - | 4 | 557 | 8861600 | ||
| BC145340.1 | - | 4 | 557 | |||

Zfp276
Genomic Map