siRNA Details
| Gene Symbol: Apba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018758.2 | NP_061228.1 | 7 | 1314 | ||
| GenBank mRNA | AF070975.1 | - | 7 | 1268 | ||
| AK045965.1 | - | 7 | 1315 | |||
| AK082046.1 | - | 1234 | ||||
| AK169462.1 | - | 7 | 1298 | |||
| AK170182.1 | - | 7 | 1262 | |||
| BC005423.1 | - | 7 | 1246 | 3487448 | ||
| BC009666.1 | - | 7 | 1258 | 3490710 | ||

Apba3
Genomic Map