siRNA Details
| Gene Symbol: Olfr1507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170918.1 | NP_001164389.1 | 2 | 1047 | ||
| NM_020512.2 | NP_065258.2 | 1 | 554 | |||
| GenBank mRNA | BC119163.1 | - | 1 | 554 | 8733912 | |
| BC119165.1 | - | 1 | 554 | 8733914 | ||

Olfr1507
Genomic Map