siRNA Details
Gene Symbol: Slc16a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020516.2 | NP_065262.1 | 5 | 1167 | ||
GenBank mRNA | BC018216.1 | - | 5 | 1167 | 4481698 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc16a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020516.2 | NP_065262.1 | 5 | 1167 | ||
GenBank mRNA | BC018216.1 | - | 5 | 1167 | 4481698 |