siRNA Details
| Gene Symbol: Srsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020587.2 | NP_065612.2 | 3 | 437 | ||
| GenBank mRNA | AK075769.1 | - | 3 | 405 | ||
| AK077648.1 | - | 3 | 423 | |||
| AK156989.1 | - | 3 | 372 | |||
| BC019437.1 | - | 3 | 397 | 4018178 | ||
| BC026944.1 | - | 3 | 1303 | 4016087 | ||
| BC040812.1 | - | 3 | 1303 | |||

Srsf4
Genomic Map