siRNA Details
| Gene Symbol: Atp5j2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020582.2 | NP_065607.1 | 2, 3 | 202 | ||
| GenBank mRNA | AB030192.1 | - | 2, 3 | 203 | ||
| AK002519.1 | - | 2, 3 | 152 | |||
| AK003235.1 | - | 2, 3 | 140 | |||
| AK003817.1 | - | 2, 3 | 152 | |||
| AK013130.1 | - | 2, 3 | 152 | |||
| AK151737.1 | - | 2, 3 | 151 | |||
| AK168095.1 | - | 2, 3 | 152 | |||
| BC029226.1 | - | 1, 2 | 142 | 2651401 | ||

Atp5j2
Genomic Map