siRNA Details
Gene Symbol: Plin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020568.3 | NP_065593.2 | 6 | 3350 | ||
GenBank mRNA | AF064748.1 | - | 6 | 3361 | ||
AK173286.1 | - | 6 | 2082 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020568.3 | NP_065593.2 | 6 | 3350 | ||
GenBank mRNA | AF064748.1 | - | 6 | 3361 | ||
AK173286.1 | - | 6 | 2082 |