siRNA Details
| Gene Symbol: March7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020575.2 | NP_065600.1 | 2 | 280 | ||
| GenBank mRNA | AF155739.1 | - | 2 | 280 | ||
| AK034109.1 | - | 305 | ||||
| AK143177.1 | - | 2 | 300 | |||
| AK166666.1 | - | 2 | 287 | |||
| AK167246.1 | - | 2 | 262 | |||
| AK168555.1 | - | 2 | 283 | |||
| BC025029.1 | - | 2 | 244 | 4935124 | ||

March7
Genomic Map