siRNA Details
| Gene Symbol: Spata5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163511.1 | NP_001156983.1 | 8 | 1482 | ||
| NM_021343.2 | NP_067318.2 | 8 | 1479 | |||
| GenBank mRNA | AF049099.1 | - | 8 | 1461 | ||
| AK011111.2 | - | 8 | 1446 | |||
| AK144998.1 | - | 8 | 1809 | |||
| BC145302.1 | - | 8 | 1450 | 9053856 | ||
| BC145309.1 | - | 8 | 1450 | |||

Spata5
Genomic Map