siRNA Details
| Gene Symbol: Tnfrsf13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021349.1 | NP_067324.1 | 3 | 414 | ||
| GenBank mRNA | AF257673.1 | - | 3 | 414 | ||
| AK155178.1 | - | 2 | 98 | |||
| BC139392.1 | - | 3 | 471 | 8862414 | ||
| BC139393.1 | - | 3 | 471 | 8862415 | ||
| BC141867.1 | - | 3 | 414 | 40054144 | ||
| BC141868.1 | - | 3 | 414 | 40054146 | ||

Tnfrsf13b
Genomic Map