siRNA Details
| Gene Symbol: Sectm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026907.3 | NP_081183.3 | 4 | 479 | ||
| GenBank mRNA | AK007321.1 | - | 3 | 341 | ||
| AK143807.1 | - | 4 | 463 | |||
| BC020159.1 | - | 3 | 430 | 4016267 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sectm1b| Gene Symbol: Sectm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026907.3 | NP_081183.3 | 4 | 479 | ||
| GenBank mRNA | AK007321.1 | - | 3 | 341 | ||
| AK143807.1 | - | 4 | 463 | |||
| BC020159.1 | - | 3 | 430 | 4016267 | ||