siRNA Details
| Gene Symbol: Srrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021403.3 | NP_067378.2 | 9 | 1208 | ||
| GenBank mRNA | BC051947.1 | - | 9 | 815 | 5696610 | |
| BC058385.1 | - | 9 | 1208 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srrm3| Gene Symbol: Srrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021403.3 | NP_067378.2 | 9 | 1208 | ||
| GenBank mRNA | BC051947.1 | - | 9 | 815 | 5696610 | |
| BC058385.1 | - | 9 | 1208 | |||