siRNA Details
| Gene Symbol: Srrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021403.3 | NP_067378.2 | 2 | 725 | ||
| GenBank mRNA | AB041661.1 | - | 2 | 367 | ||
| BC051947.1 | - | 2 | 332 | 5696610 | ||
| BC058385.1 | - | 2 | 725 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srrm3| Gene Symbol: Srrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021403.3 | NP_067378.2 | 2 | 725 | ||
| GenBank mRNA | AB041661.1 | - | 2 | 367 | ||
| BC051947.1 | - | 2 | 332 | 5696610 | ||
| BC058385.1 | - | 2 | 725 | |||