siRNA Details
| Gene Symbol: Efcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159697.1 | NP_001153169.1 | 1 | 477 | ||
| GenBank mRNA | AB041550.1 | - | 1 | 477 | ||
| BC139438.1 | - | 1 | 474 | 8862460 | ||
| BC139439.1 | - | 1 | 473 | 8862461 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efcc1| Gene Symbol: Efcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159697.1 | NP_001153169.1 | 1 | 477 | ||
| GenBank mRNA | AB041550.1 | - | 1 | 477 | ||
| BC139438.1 | - | 1 | 474 | 8862460 | ||
| BC139439.1 | - | 1 | 473 | 8862461 | ||