siRNA Details
| Gene Symbol: Fam181b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021427.2 | NP_067402.2 | 1 | 1174 | ||
| GenBank mRNA | AB041569.1 | - | 1 | 1136 | ||
| AK043957.1 | - | 1 | 1189 | |||
| AK140685.1 | - | 1 | 1127 | |||
| AK141493.1 | - | 1 | 772 | |||
| BC043329.1 | - | 1 | 1123 | 5369836 | ||
| EF537862.1 | - | 1 | 100 | |||
| EF537863.1 | - | 1 | 100 | |||

Fam181b
Genomic Map