siRNA Details
| Gene Symbol: Fam181b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021427.2 | NP_067402.2 | 1 | 948 | ||
| GenBank mRNA | AB041569.1 | - | 1 | 910 | ||
| AK043957.1 | - | 1 | 963 | |||
| AK140685.1 | - | 1 | 900 | |||
| AK141493.1 | - | 1 | 546 | |||
| BC043329.1 | - | 1 | 897 | 5369836 | ||
| Gene Symbol: Gm9934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079653.1 | - | 1 | 197 | ||

Fam181b
Genomic Map