siRNA Details
| Gene Symbol: Trim54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021447.2 | NP_067422.1 | 1 | 348 | ||
| GenBank mRNA | AF294790.1 | - | 1 | 248 | ||
| BC107258.2 | - | 1 | 93 | 40052466 | ||
| BC107259.2 | - | 1 | 93 | 40052471 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim54| Gene Symbol: Trim54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021447.2 | NP_067422.1 | 1 | 348 | ||
| GenBank mRNA | AF294790.1 | - | 1 | 248 | ||
| BC107258.2 | - | 1 | 93 | 40052466 | ||
| BC107259.2 | - | 1 | 93 | 40052471 | ||