siRNA Details
Gene Symbol: Trim54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021447.2 | NP_067422.1 | 3 | 658 | ||
GenBank mRNA | AF294790.1 | - | 3 | 558 | ||
BC107258.2 | - | 3 | 403 | 40052466 | ||
BC107259.2 | - | 3 | 403 | 40052471 |
