siRNA Details
| Gene Symbol: Pmaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021451.2 | NP_067426.1 | 2 | 258 | ||
| GenBank mRNA | AB041230.1 | - | 2 | 258 | ||
| AK043856.1 | - | 2 | 289 | |||
| AK088556.1 | - | 2 | 268 | |||
| AK143990.1 | - | 2 | 330 | |||
| AK169914.1 | - | 2 | 302 | |||
| BC050821.1 | - | 2 | 245 | 6517820 | ||

Pmaip1
Genomic Map