siRNA Details
| Gene Symbol: Kcnmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021452.1 | NP_067427.1 | 1 | 251 | ||
| GenBank mRNA | AF215892.1 | - | 1 | 251 | ||
| AK013647.1 | - | 1 | 610 | |||
| AK050596.1 | - | 1 | 151 | |||
| BC048471.1 | - | 1 | 264 | |||
| BC117779.2 | - | 1 | 251 | 40091866 | ||
| Gene Symbol: Kcnmb4os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016591.1 | - | 1 | 106 | ||

Kcnmb4
Genomic Map