siRNA Details
| Gene Symbol: Tdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021480.5 | NP_067455.5 | 6 | 882 | ||
| GenBank mRNA | AF134346.2 | - | 6 | 546 | ||
| AK077571.1 | - | 6 | 557 | |||
| AY116662.1 | - | 5 | 483 | |||
| BC058860.1 | - | 6 | 897 | 30014660 | ||

Tdh
Genomic Map