siRNA Details
| Gene Symbol: F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037085.1 | - | 9 | 900 | ||
| BC049867.1 | - | 9 | 923 | |||
| BC057921.1 | - | 9 | 911 | 5101105 | ||
| X99571.1 | - | 9 | 906 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F12| Gene Symbol: F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037085.1 | - | 9 | 900 | ||
| BC049867.1 | - | 9 | 923 | |||
| BC057921.1 | - | 9 | 911 | 5101105 | ||
| X99571.1 | - | 9 | 906 | |||