siRNA Details
| Gene Symbol: Pole3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021498.2 | NP_067473.2 | 1 | 59 | ||
| GenBank mRNA | AK007693.1 | - | 114 | |||
| BC024996.1 | - | 1 | 59 | 5253060 | ||
| BC034004.1 | - | 67 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pole3| Gene Symbol: Pole3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021498.2 | NP_067473.2 | 1 | 59 | ||
| GenBank mRNA | AK007693.1 | - | 114 | |||
| BC024996.1 | - | 1 | 59 | 5253060 | ||
| BC034004.1 | - | 67 | ||||