siRNA Details
| Gene Symbol: Med12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021521.2 | NP_067496.2 | 39 | 5841 | ||
| GenBank mRNA | AF071310.1 | - | 38 | 5591 | ||
| AK220216.1 | - | 18 | 2539 | |||
| BC057085.1 | - | 12 | 2158 | |||
| BC057119.1 | - | 38 | 5575 | |||
| BC090654.1 | - | 39 | 5839 | |||
| BC110696.1 | - | 9 | 1317 | |||
| BC145447.1 | - | 38 | 5527 | |||

Med12
Genomic Map