siRNA Details
| Gene Symbol: Huwe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021523.4 | NP_067498.4 | 69 | 11319 | ||
| GenBank mRNA | AB093227.2 | - | 35 | 6534 | ||
| AK083499.1 | - | 3 | 604 | |||
| AY772010.3 | - | 69 | 11256 | |||
| AY929611.1 | - | 67 | 10863 | |||
| BC011391.1 | - | 4 | 1018 | |||
| DQ097265.1 | - | 69 | 11275 | |||

Huwe1
Genomic Map